CAS RN 1076198-08-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Ethyl 2-[5-(2-Methyl-1,3-dioxolan-2-yl)-2-pyridyl]acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Ethyl 2-[5-(2-Methyl-1,3-dioxolan-2-yl)-2-pyridyl]acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Ethyl 2-[5-(2-Methyl-1,3-dioxolan-2-yl)-2-pyridyl]acetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.