CAS RN 107596-30-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
S-(+)-Mecamylamine Hydrochloride, TRC
CAS: 107596-30-5 Molecular Formula: C11 H21 N . Cl H Molecular Weight (g/mol): 203.75 Synonym: Bicyclo[2.2.1]heptan-2-amine, N,2,3,3-tetramethyl-, hydrochloride (1:1), (1R,2S,4S)-,Bicyclo[2.2.1]heptan-2-amine, N,2,3,3-tetramethyl-, hydrochloride, (1R,2S,4S)- (9CI),Bicyclo[2.2.1]heptan-2-amine, N,2,3,3-tetramethyl-, hydrochloride, (1R-exo)-,(+)-Mecamylamine hydrochloride,Dexmecamylamine hydrochloride,NIH 11008,NIH 11008 hydrochloride,S-(+)-Mecamylamine hydrochloride,TC 5214 hydrochloride,(S)-Mecamylamine hydrochloride IUPAC Name: (1S,3S,4R)-N,2,2,3-tetramethylbicyclo[2.2.1]heptan-3-amine;hydrochloride SMILES: Cl.CN[C@@]1(C)[C@@H]2CC[C@@H](C2)C1(C)C