CAS RN 1075727-00-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
(1R,2R)-2-(3-(tert-butoxy)-3-oxopropyl)-1-(3,4-dimethoxybenzyl)-6,7-dimethoxy-2-methyl-1,2,3,4-tetrahydroisoquinolin-2-ium benzenesulfonate (1:1), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(1R,2R)-2-(3-(tert-butoxy)-3-oxopropyl)-1-(3,4-dimethoxybenzyl)-6,7-dimethoxy-2-methyl-1,2,3,4-tetrahydroisoquinolin-2-ium benzenesulfonate (1:1), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(1R,2R)-2-(3-(tert-butoxy)-3-oxopropyl)-1-(3,4-dimethoxybenzyl)-6,7-dimethoxy-2-methyl-1,2,3,4-tetrahydroisoquinolin-2-ium benzenesulfonate (1:1), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.