CAS RN 1075-59-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4,6-Dimethoxy-1,3,5-triazin-2(1H)-one, TRC
CAS: 1075-59-8 Molecular Formula: C5 H7 N3 O3 Molecular Weight (g/mol): 157.127 Synonym: 4,6-dimethoxy-s-Triazin-2-ol,4,6-Dimethoxy-1,3,5-triazin-2(1H)-one,2-Hydroxy-4,6-dimethoxy-1,3,5-triazine,Dimethyl cyanurate IUPAC Name: 2,6-dimethoxy-1H-1,3,5-triazin-4-one SMILES: COC1=NC(=O)N=C(N1)OC