CAS RN 1073608-13-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3,4-Dehydro Cilostazol-d11, TRC
CAS: 1073608-13-5 Molecular Formula: C20H14D11N5O2 Molecular Weight (g/mol): 378.51 Synonym: 6-[4-(1-Cyclohexyl-d11-1H-tetrazol-5-yl)butoxy]-2(1H)-quinolinone,OPC 13015-d11; SMILES: O=C(N1)C=CC2=C1C=CC(OCCCCC3=NN=NN3C4([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C4([2H])[2H])=C2