CAS RN 1073-00-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Methylenecyclopropaneacetic Acid, TRC
CAS: 1073-00-3 Molecular Formula: C6 H8 O2 Molecular Weight (g/mol): 112.126 Synonym: (Methylenecyclopropyl)acetic Acid,MCPA IUPAC Name: 2-(2-methylenecyclopropyl)acetic acid SMILES: OC(=O)CC1CC1=C