CAS RN 1072902-84-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
(3S,4S)-3-Hexyl-4[(S)-2-(triisopropylsilyloxy)tridecyl]-2-oxetanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(3S,4S)-3-Hexyl-4[(S)-2-(triisopropylsilyloxy)tridecyl]-2-oxetanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(3S,4S)-3-Hexyl-4[(S)-2-(triisopropylsilyloxy)tridecyl]-2-oxetanone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.