accessibility menu, dialog, popup

UserName

H3CNCH3H3COOH

CAS RN 1072069-91-0

IUPAC Name: 3-(2,3,3-trimethyl-3H-indol-5-yl)propanoic acid
Synonyms: 3-(2,3,3-trimethylindol-5-yl)propanoic acid
Molecular Weight (g/mol): 231.3
Molecular Formula: C14H17NO2
InChi Key: QIMIOCZMNPBLPO-UHFFFAOYSA-N
SMILES: CC1=NC2=C(C=C(CCC(O)=O)C=C2)C1(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results