CAS RN 1071929-03-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
DL-Dapoxetine Hydrochloride, TRC
CAS: 1071929-03-7 Molecular Formula: C21 H23 N O . Cl H Molecular Weight (g/mol): 341.874 Synonym: Benzenemethanamine, N,N-dimethyl-α-[2-(1-naphthalenyloxy)ethyl]-, hydrochloride (1:1) (ACI),N,N-Dimethyl-α-[2-(1-naphthalenyloxy)ethyl]benzenemethanamine hydrochloride,(±)-Dapoxetine hydrochloride,N,N-Dimethyl-3-(1-naphthoxy)-1-phenyl-1-propylamine hyd IUPAC Name: N,N-dimethyl-3-naphthalen-1-yloxy-1-phenylpropan-1-amine;hydrochloride SMILES: Cl.CN(C)C(CCOc1cccc2ccccc12)c3ccccc3