CAS RN 1070663-78-3
IUPAC Name:
dicyclohexyl[3,6-dimethoxy-2',4',6'-tris(propan-2-yl)-[1,1'-biphenyl]-2-yl]phosphane
Synonyms:
dicyclohexyl({2',4',6'-triisopropyl-3,6-dimethoxy-[1,1'-biphenyl]-2-yl})phosphane
Molecular Weight (g/mol):
536.78
Molecular Formula:
C35H53O2P
InChi Key:
WDVGNXKCFBOKDF-UHFFFAOYSA-N
SMILES:
COC1=C(P(C2CCCCC2)C2CCCCC2)C(=C(OC)C=C1)C1=C(C=C(C=C1C(C)C)C(C)C)C(C)C
1
–
3
of
3
results
1
BrettPhos, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
BrettPhos, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
BrettPhos, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.