CAS RN 1070377-34-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-[[6-Chloro-2-[(4-cyanophenyl)amino]-4-pyrimidinyl]oxy]-3,5-dimethylbenzonitrile (Etravirine Impurity 23), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.