CAS RN 106931-78-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fluocinolone Acetonide-21-carboxylic Acid (>90%), TRC
CAS: 106931-78-6 Molecular Formula: C24 H28 F2 O7 Molecular Weight (g/mol): 466.47 Synonym: Pregna-1,4-dien-21-oic acid, 6,9-difluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]-3,20-dioxo-, (6α,11β,16α,17α)- (9CI),6bH-Naphth[2',1':4,5]indeno[1,2-d][1,3]dioxole, pregna-1,4-dien-21-oic acid deriv.,6α,9-Difluoro-11β-hydroxy-16α,17-(1-methylethylidenedioxy)-3,20-dioxopregna-1,4-dien-21-oic acid,Fluocinolone Acetonide 21-carboxylic acid SMILES: CC1(C)O[C@@H]2C[C@H]3[C@@H]4C[C@H](F)C5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)C(=O)O