CAS RN 106686-60-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Ethyl(1-methylbutyl)malonuric Acid, TRC
CAS: 106686-60-6 Molecular Formula: C11H20N2O4 Molecular Weight (g/mol): 244.29 Synonym: 2-[[(Aminocarbonyl)amino]carbonyl]-2-ethyl-3-methylhexanoic Acid,N-Carbamoyl-2-ethyl-2-(1-methylbutyl)malonamic Acid; IUPAC Name: 2-(carbamoylcarbamoyl)-2-ethyl-3-methylhexanoic acid SMILES: CCCC(C)C(C(NC(N)=O)=O)(C(O)=O)CC