CAS RN 1062-64-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
(11Beta,16Alpha)-9-Fluoro-11-hydroxy-17,21-[(1-methoxypentylidene)bis(oxy)]-16-methyl-pregna-1,4-diene-3,20-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(11Beta,16Alpha)-9-Fluoro-11-hydroxy-17,21-[(1-methoxypentylidene)bis(oxy)]-16-methyl-pregna-1,4-diene-3,20-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(11Beta,16Alpha)-9-Fluoro-11-hydroxy-17,21-[(1-methoxypentylidene)bis(oxy)]-16-methyl-pregna-1,4-diene-3,20-dione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.