CAS RN 105990-27-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-(-)-Higenamine Hydrobromide, TRC
CAS: 105990-27-0 Molecular Formula: C16H18BrNO3 Molecular Weight (g/mol): 352.22 Synonym: 1,2,3,4-Tetrahydro-1-[(4-hydroxyphenyl)methyl]-6,7-isoquinolinediol,1,2,3,4-Tetrahydro-1-[(4-hydroxyphenyl)methyl]-hydrobromide-(1S)-6,7-isoquinolinediol,(S)-Hydrobromide,(-)-Higenamine Hydrobromide,(S)-(-)-Norcoclaurine Hydrobromide IUPAC Name: (1S)-1-[(4-hydroxyphenyl)methyl]-1,2,3,4-tetrahydroisoquinoline-6,7-diol;hydrobromide SMILES: Br.Oc1ccc(C[C@@H]2NCCc3cc(O)c(O)cc23)cc1