CAS RN 105959-56-6
CAS RN 105959-56-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
N-(alpha-Phenylethyl)ibuprofenamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
N-(1-Phenylethyl) Ibuprofen Amide(Mixture of 4 Diastereomers), TRC
CAS: 105959-56-6 Molecular Formula: C21 H27 N O Molecular Weight (g/mol): 309.45 Synonym: Benzeneacetamide, α-methyl-4-(2-methylpropyl)-N-(1-phenylethyl)-,2-(4-isobutylphenyl)-N-(1-phenylethyl)propanamide,N-(α-Phenylethyl)ibuprofenamide IUPAC Name: 2-[4-(2-methylpropyl)phenyl]-N-(1-phenylethyl)propanamide SMILES: CC(C)Cc1ccc(cc1)C(C)C(=O)NC(C)c2ccccc2