CAS RN 1059188-84-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
(3R,5R)-5-(3-Hydroxyphenyl)-3-[[(1R)-1-phenylethyl]amino]-1-[4-(trifluoromethyl)phenyl]-2-pyrrolidinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(3R,5R)-5-(3-Hydroxyphenyl)-3-[[(1R)-1-phenylethyl]amino]-1-[4-(trifluoromethyl)phenyl]-2-pyrrolidinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(3R,5R)-5-(3-Hydroxyphenyl)-3-[[(1R)-1-phenylethyl]amino]-1-[4-(trifluoromethyl)phenyl]-2-pyrrolidinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.