CAS RN 105675-85-2
IUPAC Name:
4-bromo-1-methyl-1H-pyrazol-5-amine
Synonyms:
4-bromo-2-methylpyrazol-3-amine
Molecular Weight (g/mol):
176.02
Molecular Formula:
C4H6BrN3
InChi Key:
ODFDZOQJRPDQDF-UHFFFAOYSA-N
SMILES:
CN1N=CC(Br)=C1N
1
–
3
of
3
results
1
3-Amino-4-bromo-2-methylpyrazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
3-Amino-4-bromo-2-methylpyrazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3-Amino-4-bromo-2-methylpyrazole, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.