CAS RN 105562-12-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(11Beta,17E)-11,20-dihydroxy-3-oxo-pregna-4,17(20)-dien-21-al, TRC
CAS: 105562-12-7 Molecular Formula: C21H28O4 Molecular Weight (g/mol): 344.44 Synonym: Pregna-4,17(20)-dien-21-al, 11,20-dihydroxy-3-oxo-, (11β,17E)- (9CI, ACI),(11β,17E)-11,20-Dihydroxy-3-oxopregna-4,17(20)-dien-21-al (ACI),(17E)-11β,20-Dihydroxy-3-oxopregna-4,17(20)-dien-21-al IUPAC Name: (2E)-2-hydroxy-2-[(8S,9S,10R,11S,13S,14S)-11-hydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-ylidene]acetaldehyde SMILES: C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC/C/2=C(\O)/C=O