CAS RN 10545-23-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-[(Dimethylamino)methyl]-2-hexanone, TRC
CAS: 10545-23-0 Molecular Formula: C9H19NO Molecular Weight (g/mol): 157.25 IUPAC Name: 3-[(dimethylamino)methyl]hexan-2-one SMILES: CCCC(CN(C)C)C(=O)C