CAS RN 1051389-99-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac 1,2-Bislauroyl-3-chloropropanediol, TRC
CAS: 1051389-99-1 Molecular Formula: C27H51ClO4 Molecular Weight (g/mol): 475.14 Synonym: 1,1'-[1-(Chloromethyl)-1,2-ethanediyl] Ester Dodecanoic Acid IUPAC Name: [(2S)-3-chloro-2-dodecanoyloxypropyl] dodecanoate SMILES: O=C(CCCCCCCCCCC)OC[C@H](OC(CCCCCCCCCCC)=O)CCl