CAS RN 105091-67-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Niobium n-pentyloxide, 99% (metals basis), Thermo Scientific™
CAS: 105091-67-6 MDL Number: MFCD00144918