CAS RN 1049733-10-9
IUPAC Name:
2-(prop-2-yn-1-yl)pyrrolidine-2-carboxylic acid hydrochloride
Synonyms:
2-(prop-2-yn-1-yl)pyrrolidine-2-carboxylic acid hydrochloride
Molecular Weight (g/mol):
189.64
Molecular Formula:
C8H12ClNO2
InChi Key:
HUPSJXIFWSWGFS-UHFFFAOYNA-N
SMILES:
Cl.OC(=O)C1(CC#C)CCCN1
1
–
3
of
3
results
1
(R)-Alpha-Propynyl-proline Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(R)-a-Propynyl-proline Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(R)-Alpha-Propynyl-proline Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.