CAS RN 10482-56-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-Alpha-Terpineol, TRC
CAS: 10482-56-1 Molecular Formula: C10 H18 O Molecular Weight (g/mol): 154.25 Synonym: 3-Cyclohexene-1-methanol, α,α,4-trimethyl-, (1S)-,3-Cyclohexene-1-methanol, α,α,4-trimethyl-, (S)-,p-Menth-1-en-8-ol, (S)-(-)- (8CI),(1S)-α,α,4-Trimethyl-3-cyclohexene-1-methanol,(-)-α-Terpineol,(S)-(-)-Terpineol,(S)-(-)-α-Terpineol,(S)-α,α,4-Trimethyl-3-cyclohexene-1-methanol,(S)-α-Terpineol,l-α-Terpineol,α-Terpineol, (-)-,(S)-alpha-Terpineol IUPAC Name: 2-[(1S)-4-methylcyclohex-3-en-1-yl]propan-2-ol SMILES: CC1=CC[C@H](CC1)C(C)(C)O