CAS RN 1047753-79-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
4-(1,3-Benzodioxol-5-yl)-1,4,6,7-tetrahydro-1-methyl-7-oxo-3-phenyl-8H-pyrazolo[3,4-e][1,4]thiazepine-8-acetic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
4-(1,3-Benzodioxol-5-yl)-1,4,6,7-tetrahydro-1-methyl-7-oxo-3-phenyl-8H-pyrazolo[3,4-e][1,4]thiazepine-8-acetic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4-(1,3-Benzodioxol-5-yl)-1,4,6,7-tetrahydro-1-methyl-7-oxo-3-phenyl-8H-pyrazolo[3,4-e][1,4]thiazepine-8-acetic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.