CAS RN 104700-12-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,2-Ethylene Glycol-13C2, TRC
CAS: 104700-12-1 Molecular Formula: 13C2H6O2 Molecular Weight (g/mol): 64.05 Synonym: 1,2-Ethanediol-13C2 IUPAC Name: (1,2-13C2)ethane-1,2-diol SMILES: O[13C]([H])([H])[13C]([H])([H])O