CAS RN 104222-42-6
CAS RN 104222-42-6
IUPAC Name:
3-bromo-2,4,5-trifluorobenzoic acid
Synonyms:
3-bromo-2,4,5-trifluorobenzoic acid
Molecular Weight (g/mol):
254.99
Molecular Formula:
C7H2BrF3O2
InChi Key:
VWVLSESGZBAPHA-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC(F)=C(F)C(Br)=C1F
1
–
1
of
1
results
1
3-Bromo-2,4,5-trifluorobenzoic acid, 98%
CAS: 104222-42-6 Molecular Formula: C7H2BrF3O2 Molecular Weight (g/mol): 254.99 MDL Number: MFCD05664065 InChI Key: VWVLSESGZBAPHA-UHFFFAOYSA-N Synonym: 2,4,5-trifluoro-3-bromobenzoic acid,benzoic acid, 3-bromo-2,4,5-trifluoro,3-bromanyl-2,4,5-tris fluoranyl benzoic acid,pubchem1366,3-bromo-2,4,5-trifluoro-benzoic acid,acmc-20an96,3-bromo-2,4,5-trifluoro benzoic acid PubChem CID: 2783246 IUPAC Name: 3-bromo-2,4,5-trifluorobenzoic acid SMILES: C1=C(C(=C(C(=C1F)F)Br)F)C(=O)O