CAS RN 104-27-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Alpha-Methylanisalacetone, TRC
CAS: 104-27-8 Molecular Formula: C12 H14 O2 Molecular Weight (g/mol): 190.2384 Synonym: 1-Penten-3-one, 1-(p-methoxyphenyl)- (6CI,7CI,8CI),1-(4-Methoxyphenyl)-1-penten-3-one,1-p-Methoxyphenylpent-1-en-3-one,Ethyl p-methoxystyryl ketone,NSC 133448,α-Methylanisalacetone IUPAC Name: (E)-1-(4-methoxyphenyl)pent-1-en-3-one SMILES: CCC(=O)\C=C\c1ccc(OC)cc1