CAS RN 103914-79-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Oxo-1,4-dihydroquinoline-3-carboxamide, TRC
CAS: 103914-79-0 Molecular Formula: C10H8N2O2 Molecular Weight (g/mol): 188.18 IUPAC Name: 4-oxo-1H-quinoline-3-carboxamide SMILES: NC(=O)C1=CNc2ccccc2C1=O