CAS RN 103877-02-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-O-Desmethyl Omeprazole Sulfide, TRC
CAS: 103877-02-7 Molecular Formula: C16H17N3O2S Molecular Weight (g/mol): 315.39 Synonym: 1H-Benzimidazol-6-ol, 2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]thio]-,2-[(4-methoxy-3,5-dimethyl-2-pyridyl)methylsulfanyl]-1H-benzimidazol-5-ol SMILES: COc1c(C)cnc(CSc2nc3cc(O)ccc3[nH]2)c1C