accessibility menu, dialog, popup

UserName

CAS RN 1036991-25-9

BOOH3CCH3H3CH3CONO

CAS RN 1036991-25-9

IUPAC Name: 4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]morpholine
Synonyms: 4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoyl]morpholine
Molecular Weight (g/mol): 317.19
Molecular Formula: C17H24BNO4
InChi Key: PKONGXZPUDFPPH-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(=CC=C1)C(=O)N1CCOCC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results