CAS RN 1034298-00-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Hydroxy-4-methyl-17-ethynyl-3-dehydroxyestradiol, TRC
CAS: 1034298-00-4 Molecular Formula: C21H26O2 Molecular Weight (g/mol): 310.43 Synonym: (8S,9S,13S,14S,17R)-17-Ethynyl-4,13-dimethyl-7,8,9,11,12,13,14,15,16,17-decahydro-6H-cyclopenta[a]phenanthrene-1,17-diol IUPAC Name: (8S,9S,13R,14S,17R)-17-ethynyl-4,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthrene-1,17-diol SMILES: Cc1ccc(O)c2[C@H]3CC[C@]4(C)[C@@H](CC[C@@]4(O)C#C)[C@@H]3CCc12