CAS RN 103254-49-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Cyclohexylpropan-1-amine, TRC
CAS: 103254-49-5 Molecular Formula: C9H19N Molecular Weight (g/mol): 141.25 IUPAC Name: 2-cyclohexylpropan-1-amine SMILES: CC(CN)C1CCCCC1