CAS RN 103201-79-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(3R,7aS)-Tetrahydro-3-phenyl-3H,5H-pyrrolo[1,2-c]oxazol-5-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(3R,7aS)-Tetrahydro-3-phenyl-3H,5H-pyrrolo[1,2-c]oxazol-5-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.