accessibility menu, dialog, popup

UserName

FH2NClCl

CAS RN 1017777-51-3

IUPAC Name: 2,3-dichloro-6-fluoroaniline
Synonyms: 2,3-dichloro-6-fluoroaniline
Molecular Weight (g/mol): 180
Molecular Formula: C6H4Cl2FN
InChi Key: WBHSLAUXVJCSBO-UHFFFAOYSA-N
SMILES: NC1=C(F)C=CC(Cl)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results