accessibility menu, dialog, popup

UserName

CAS RN 101-29-1

OHONIOI

CAS RN 101-29-1

IUPAC Name: 2-(3,5-diiodo-4-oxo-1,4-dihydropyridin-1-yl)acetic acid
Synonyms: iodopyracet 碘奥酮 diodone
Molecular Weight (g/mol): 404.93
Molecular Formula: C7H5I2NO3
InChi Key: PVBALTLWZVEAIO-UHFFFAOYSA-N
SMILES: OC(=O)CN1C=C(I)C(=O)C(I)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results