accessibility menu, dialog, popup

UserName

CH3BOHHONOO

CAS RN 100960-11-0

IUPAC Name: (2-methyl-5-nitrophenyl)boronic acid
Synonyms: 2-methyl-5-nitrophenylboronic acid
Molecular Weight (g/mol): 180.95
Molecular Formula: C7H8BNO4
InChi Key: HRJFQIRIDFXMNX-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1B(O)O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results