missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Diazomethane Generators for Gas Chromatography
For generating diazomethane and other gases for gas chromatography, mass sepctroscopy and NMR studies
£157.00 - £180.00
Specifications
Certifications/Compliance | USP Type I, ASTM E 438 Type I, Class A |
---|---|
Type | Diazomethane Generator |
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
12418922
|
DWK Life Sciences
281155 |
20 cm |
£180.00
Each |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12408922
|
DWK Life Sciences
281135 |
19 cm |
£157.00
Each |
Estimated Shipment: 05-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Utilizes clear-seal grease-free joints
- «O« ring not required
- Manufactured from low extractable borosilicate glass that conforms to USP Type I and ASMT™ E 438 Type I, Class A requirements
Specifications
USP Type I, ASTM E 438 Type I, Class A | |
Diazomethane Generator |