Learn More
DWK Life Sciences Wheaton™ Diazomethane Generators for Gas Chromatography
For generating diazomethane and other gases for gas chromatography, mass sepctroscopy and NMR studies
Brand: DWK Life Sciences 281135
Additional Details : Weight : 0.36290kg
Description
- Utilizes clear-seal grease-free joints
- «O« ring not required
- Manufactured from low extractable borosilicate glass that conforms to USP Type I and ASMT™ E 438 Type I, Class A requirements
Specifications
USP Type I, ASTM E 438 Type I, Class A | |
19 cm | |
Diazomethane Generator |
7.4 in. | |
Plastic standard taper joint clamp, a 13-425 screw cap with cut-out top and a ptfe faced rubber liner |