missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pro-Lab Diagnostics™ Microbank™ Freezer Storage Boxes
Suitable for collection and organization in low temperature freezers.
£59.40 - £63.65
Product Code | Brand | Closure Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Closure Color | Price | Quantity & Availability | |||||
12807775
|
Pro-Lab Diagnostics™
PL.169/B-1 |
Blue |
£63.65
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12817775
|
Pro-Lab Diagnostics™
PL.169/R-1 |
Red |
£59.40
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
Description
- Manufactured from durable plastic
- Each freezer box holds up to 81 Microbank vials
- With “see through” lid, number locator printed screen, and tube collection device
- Available in blue or red
- Dimensions: 133 × 133 × 55mm
Specifications
Each | |
13.3 cm | |
5.23 in. |