Learn More
Thermo Scientific™ Orion™ AQUAfast™ AQ3170 Colorimeter
Gather accurate chlorine measurements with a colorimeter designed for both the laboratory and the field in markets such as drinking water, wastewater, food & beverage, aquaculture, and agriculture.
£33.90 - £495.64
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15971352
|
Thermo Scientific™
AQ3170 |
AQUAfast AQ3170 Chlorine Colorimeter |
£495.64
Each |
Estimated Shipment: 11-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15984002
|
Thermo Scientific™
CLSK100 |
AQUAfast chlorine primary standard kit to prepare 1.5 mg/L NIST-traceable chlorine standard |
£33.90
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15994002
|
Thermo Scientific™
CLSK200 |
AQUAfast chlorine secondary standards (0.0, 0.20 and 1.0 mg/L chlorine) for AQ3170 colorimeter |
£152.31
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15904002
|
Thermo Scientific™
AC2V10 |
AQUAfast 10mm short path-length plastic vials for high level chlorine, 12 pack |
£149.72
Each |
Estimated Shipment: 22-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Thermo Scientific™ Orion™ AQUAfast™ AQ3170 colorimeter has a rugged and waterproof design to provide accurate chlorine measurements whether testing in the laboratory or the field. This colorimeter is designed to improve the user experience by including a large, backlit display, a real time clock and automatic data logging. The AQUAfast AQ3170 colorimeter is used in markets such as drinking water, wastewater, food & beverage, aquaculture, and agriculture.
The Orion AQUAfast AQ3170 chlorine colorimeter is ideal for the dedicated free and total chlorine testing of water and wastewater and is designed to assist with US EPA testing compliance for free and total chlorine measurements of drinking water and wastewater samples. Using an LED light source at 530 nm, this colorimeter measures two free and total chlorine testing ranges of 0.02 to 2.00mg/L chlorine using powder DPD reagents and standard 24mm vials and 0.1 to 8.0mg/L chlorine using powder DPD reagents and new 10mm plastic vials. The AQUAfast AQ3170 chlorine colorimeter is CE certified, IP-67 waterproof and accepts Orion AQUAfast free and total chlorine powder DPD reagents, available as standard 100 individual powder packs and new bulk powder dispensers with refillable vials for 250 dispense counts each. The Orion AQUAfast primary and secondary chlorine standards are also now available as separate accessories for the AQUAfast AQ3170 chlorine colorimeter.- Convenient scroll-driven menu system
- Automatic switch-off
- 16-point internal automatic data storage with date/time stamp
- Easy-to-read, backlit display
- One time zero function
- Waterproof design
- Angled, ergonomic design for easy bench top or hand-held use
- Calibration to government or organizational standards (ex. NIST or ISO)
- 2 year warranty
Warranty
2 years