Learn More
Thermo Scientific™ Orion™ AQUAfast™ AQ3170 Colorimeter
Gather accurate chlorine measurements with a colorimeter designed for both the laboratory and the field in markets such as drinking water, wastewater, food & beverage, aquaculture, and agriculture.
Brand: Thermo Scientific™ AQ3170
Code : NEW
Additional Details : Weight : 0.26000kg
Description
The Thermo Scientific™ Orion™ AQUAfast™ AQ3170 colorimeter has a rugged and waterproof design to provide accurate chlorine measurements whether testing in the laboratory or the field. This colorimeter is designed to improve the user experience by including a large, backlit display, a real time clock and automatic data logging. The AQUAfast AQ3170 colorimeter is used in markets such as drinking water, wastewater, food & beverage, aquaculture, and agriculture.
- Convenient scroll-driven menu system
- Automatic switch-off
- 16 point internal automatic data storage with date/time stamp
- Easy to read, backlit display
- One time zero function
- Waterproof design
- Angled, ergonomic design for easy bench top or hand-held use
- Calibration to government or organizational standards (ex. NIST or ISO)
- Calibration can be refined for very specific measurement ranges
- Account for any interferences that are known to be present in the sample
- Ability to export data to a PC
- 2-year warranty
Services
2 years
Specifications
±0.05 mg/L (0.02 to 1.00 mg/L); ±0.10 mg/L (1.01 to 2.00 mg/L); ±0.20 mg/L (2.01 to 3.00 mg/L); ±0.30 mg/L (3.01 to 4.00 mg/L); ±0.40 mg/L (4.01 to 8.00 mg/L) | |
17 hours operating time or 5,000 measurements in continuous mode when display backlight is off | |
CE | |
AQUAfast AQ3170 Chlorine Colorimeter | |
Backlit LCD | |
IP67 | |
AQUAfast | |
Reagents: AC4P71, AC4P72, AQ250F, AQ250T; Vials: AC2V24, AC2V10; Standards: CLSK100, CLSK200 | |
Powder packs, bulk powder dispensers | |
Dedicated Chlorine Colorimeter | |
530 nm | |
260 g (With batteries) |
Yes | |
User and factory calibration functions | |
16 with date/time stamp | |
4 x AAA | |
155 x 75 x 35 mm | |
LED | |
Chlorine Colorimeter | |
Colorimeter with free and total chlorine powder packs (100 each), sample vials with lids, field case, batteries and literature | |
Low Range: 0.02 to 2.00 mg/L; High Range: 0.10 to 8.00 mg/L | |
2 Years | |
± 1 nm |