missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MULTIROIR Euronorm ESD Container
Simplified cleaning thanks to smooth internal walls designed for the storage and transport of electronic products
Brand: MULTIROIR AESDEG64/17HG
Additional Details : Weight : 1.84000kg
Description
- Euronorm ESD container
- Material: polypropylene 2 integrated ergonomic handles
- Simplified cleaning thanks to smooth internal walls
- Specially designed for the storage and transport of electronic products
Specifications
Storage Box | |
Gray | |
BP6170 K |
Polypropylene | |
Rectangular | |
600 x 400 x 170 mm |