missing translation for 'onlineSavingsMsg'
Learn More

MULTIROIR Euronorm ESD Container

Simplified cleaning thanks to smooth internal walls designed for the storage and transport of electronic products

Brand:  MULTIROIR AESDEG86/42HG

Additional Details : Weight : 6.93000kg

 View more versions of this product

Product Code. 17206880

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Euronorm ESD container
  • Material: polypropylene 2 integrated ergonomic handles
  • Simplified cleaning thanks to smooth internal walls
  • Specially designed for the storage and transport of electronic products
Specifications

Specifications

Storage Box
Gray
BP8420 K
Polypropylene
Rectangular
800 x 600 x 420 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers