Learn More
Thermo Scientific™ ES Series Lab Freezers
Save valuable space with Thermo Scientific™ ES Series lab freezers, designed for general-purpose lab storage and routine sample protection.
£997.00
Products 1
Product Code Brand Capacity (Metric) Plug Type Price Quantity & Availability  
Product Code Brand Capacity (Metric) Plug Type Price Quantity & Availability  
16610223
missing translation for 'viewDocuments' Promotion Details
Thermo Scientific™
98F-AEV-TS
98 L EU
£997.00
Each
View alternative products
missing translation for 'toSaveUpto' 
»
In Stock
to see stock.
Description
Description
Save valuable space with Thermo Scientific™ ES Series Lab Freezers, designed for general-purpose lab storage and routine sample protection.
Features:
  • Integrated controller
  • Digital display
  • High- and low-temperature alarms
  • Standard door locks
  • Low energy consumption
  • Access ports
  • CE marked
Recommended for:
General-purpose lab storage
Ordering Information:
Available in 50Hz regions only; not available in North America or other 60Hz regions.
  • Warranty varies by country, please contact us for details
  • Videos