Learn More
Alfa Aesar™ Zymosan, Saccharomyces cerevisiae
Brand:  Alfa Aesar™ J62876.MD
Code : NEW
Additional Details : CAS Number : 58856-93-2 Weight : 0.00025kg
Quantity 250mg
 View more versions of this product
Product Code. 15455179
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
Description
Description
An insoluble preparation of yeast cell, has been shown to activate macrophages via toll-like receptor 2.
Specifications
Chemical Identifiers
Zymosan
Saccharomyces cerevisiae
14,10200
58856-93-2
MFCD00082157
Specifications
Zymosan
58856-93-2
White
250mg
Saccharomyces cerevisiae
Powder
MFCD00082157
14,10200