missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Zymogram Developing Buffer, 10X, pH 7.45

Brand:  Alfa Aesar™ J61375.AP

Code : NEW

Additional Details : Weight : 0.00050kg

 View more versions of this product

Product Code. 15443979

  • £22.50 / 500mL


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Specifications

Colorless
Viscous Liquid
2° to 8°C
Odorless
500mL
10X
Buffer
Videos
SDS