missing translation for 'onlineSavingsMsg'
Learn More

Zotepine, Tocris Bioscience™

5-HT2A/D2 antagonist; atypical antipsychotic

Brand:  Tocris (Bio-Techne) 399610

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13463944

  • £103.95 / 10mg


Estimated Shipment
04-04-2019
Add to basket

Description

Description

5-HT 2A receptor and dopamine D 2 receptor antagonist (K i values are 0.69 and 2.3 nM respectively). Also a potent histamine H 1 receptor antagonist (IC 50 = 8.0 nM). Atypical antipsychotic; exhibits antidepressive and anxiolytic effects in vivo.
Specifications

Chemical Identifiers

26615-21-4
331.858
zotepine, nipolept, lodopin, zoleptil, compound-4, zotepina, zotepinum, zotepina spanish, zotepinum inn-latin, zotepina inn-spanish
CHEBI:32316
CN(C)CCOC1=CC2=CC=CC=C2SC3=C1C=C(C=C3)Cl
C18H18ClNOS
HDOZVRUNCMBHFH-UHFFFAOYSA-N
5736
2-(3-chlorobenzo[b][1]benzothiepin-5-yl)oxy-N,N-dimethylethanamine

Specifications

Zotepine
26615-21-4
zotepine, nipolept, lodopin, zoleptil, compound-4, zotepina, zotepinum, zotepina spanish, zotepinum inn-latin, zotepina inn-spanish
CN(C)CCOC1=CC2=CC=CC=C2SC3=C1C=C(C=C3)Cl
331.858
CHEBI:32316
>99%
10mg
C18H18ClNOS
HDOZVRUNCMBHFH-UHFFFAOYSA-N
2-(3-chlorobenzo[b][1]benzothiepin-5-yl)oxy-N,N-dimethylethanamine
5736
331.86
Videos