Learn More
Zonisamide, Tocris Bioscience™
Blocks voltage-sensitive Na+ and Ca2+ channels (T-type)
Brand:  Tocris (Bio-Techne) 262550
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13441594
  • £257.41 / 50mg
Estimated Shipment
28-06-2019
Description
Description
Antiepileptic that possesses a broad spectrum anticonvulsant and mechanistic profile. Blocks voltage-sensitive Na+ and T-type Ca2+ channels, stimulates BKCa channels, modulates GABA, glutamate and monoamine neurotransmission, inhibits lipid peroxidation and scavenges hydroxyl and nitric oxide free radicals. Displays neuroprotective and antiParkinsonian activity.
Specifications
Chemical Identifiers
68291-97-4
212.223
zonisamide, zonegran, exceglan, excegram, 1,2-benzisoxazole-3-methanesulfonamide, zonisamida, zonisamidum, zonisamidum latin, zonisamida spanish, excegran
CHEBI:10127
C1=CC=C2C(=C1)C(=NO2)CS(=O)(=O)N
C8H8N2O3S
UBQNRHZMVUUOMG-UHFFFAOYSA-N
5734
1,2-benzoxazol-3-ylmethanesulfonamide
Specifications
Zonisamide
68291-97-4
zonisamide, zonegran, exceglan, excegram, 1,2-benzisoxazole-3-methanesulfonamide, zonisamida, zonisamidum, zonisamidum latin, zonisamida spanish, excegran
C1=CC=C2C(=C1)C(=NO2)CS(=O)(=O)N
212.223
CHEBI:10127
>99%
50mg
C8H8N2O3S
UBQNRHZMVUUOMG-UHFFFAOYSA-N
1,2-benzoxazol-3-ylmethanesulfonamide
5734
212.22
Videos