missing translation for 'onlineSavingsMsg'
Learn More

Zolantidine dimaleate, Tocris Bioscience™

Potent, centrally active H2 antagonist

Brand:  Tocris (Bio-Techne) 107010

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13438443

  • £160.23 / 10mg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A potent, selective and brain penetrating H2 receptor antagonist.
Specifications

Chemical Identifiers

104076-39-3
617.714
N-[3-[3-(1-Piperidinylmethyl)phenoxy]propyl]-2-benzothiazolamine dimaleate
formic acid;N-[3-[3-(piperidin-1-ylmethyl)phenoxy]propyl]-1,3-benzothiazol-2-amine;prop-2-enoic acid
C30H39N3O9S
LUWINHZYJPMGDV-UHFFFAOYSA-N
77519075
C=CC(=O)O.C=CC(=O)O.C1CCN(CC1)CC2=CC(=CC=C2)OCCCNC3=NC4=CC=CC=C4S3.C(=O)O.C(=O)O

Specifications

Zolantidine dimaleate
104076-39-3
N-[3-[3-(1-Piperidinylmethyl)phenoxy]propyl]-2-benzothiazolamine dimaleate
C=CC(=O)O.C=CC(=O)O.C1CCN(CC1)CC2=CC(=CC=C2)OCCCNC3=NC4=CC=CC=C4S3.C(=O)O.C(=O)O
617.714
613.68
10mg
C30H39N3O9S
LUWINHZYJPMGDV-UHFFFAOYSA-N
formic acid;N-[3-[3-(piperidin-1-ylmethyl)phenoxy]propyl]-1,3-benzothiazol-2-amine;prop-2-enoic acid
77519075
>98%
Videos
Documents